Learn More
Gibco™ Penicillin-Streptomycin (5,000 U/mL)
For prevention from bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
Supplier: Gibco™ 15070063
Description
Penicillin was originally purified from fungi Penicillium and acts by interfering directly with turnover of bacterial cell wall and indirectly by triggering release of enzymes that alter cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of the bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.
- Solution contains 5000 units/mL of penicillin and 5000μg/mL of streptomycin
Cell Culture, Mammalian Cell Culture, Primary Cell Culture, T Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: -5°C to -20°C Shipping conditions: Dry ice Shelf life: 12 months from date of manufacture |
|
Mammalian Cell Culture, Insect Cell Culture | |
Antibiotic | |
Prevention of Cell Culture Contamination | |
Dry Ice |
100 X | |
Liquid | |
Sterile-filtered | |
100 mL |
For Research Use Only. Not for use in diagnostic procedures.