Learn More
Gibco™ Poloxamer 188 Non-ionic Surfactant (10%)
Poloxamer 188 is a non-ionic surfactant used to control shear forces in suspension cultures, reduce foaming in stirred cultures, and minimize cell attachment to glass surfaces. This product comes as a 10% solution, sterile-filtered and ready for use in cell culture applications.
Supplier: Gibco™ 24040032
Description
Gibco Poloxamer 188 is a non-ionic surfactant used to control shear forces in suspension cultures, reduce foaming in stirred cultures, and minimize cell attachment to glass surfaces. It is widely utilized to protect cells in suspension from damage during processes such as transfer, freezing, thawing, and stirring. Its ability to stabilize cell membranes and reduce surface tension helps cell viability and integrity, particularly in mammalian cell cultures, including translational and clinical research applications.
Key features
- Animal origin-free—suitable for applications requiring non-animal derived components
- Sterile-filtered solution—ready-for-use in your cell culture applications
- Manufacturing quality—cGMP-compliant under the ISO 13485 standard
Why choose Poloxamer 188 Non-ionic Surfactant (10%)
- Proven track record—Gibco products are renowned for their quality and reliability, backed by extensive research and a strong publication record
- Quality assurance—each lot of Poloxamer 188 is rigorously tested to ensure it meets Gibco internal specifications for supporting cell growth and viability
- Expert support—Thermo Fisher Scientific provides comprehensive technical support and resources to assist with your cell culture needs
- Wide range of applications—suitable for various cell culture applications, including protein expression, virus production, and more
This high-quality Poloxamer 188 formulation, combined with the Gibco commitment to excellence, ensures reliable and reproducible results in your cell culture experiments. Choose Gibco products for your cell culture needs and benefit from our decades of expertise and innovation in the field.
Additional information
- Concentration: provided at 10%, effective at a working concentration of 0.1%
- Osmolality: 65–105 mOsm/kg
- pH: 5.0–7.5
- Sterility: sterile-filtered, tested using current edition of USP
- Performance assay: each lot tested for ability to support growth of 293-F cells in FreeStyle 293 Expression Medium
- Form: liquid
Order Info
Shipping Condition: Room Temperature
Specifications
| Storage conditions: 15 – 30°C Shipping conditions: Room temperature Shelf life: 18 months from date of manufacture |
|
| Animal Origin-free | |
| Suspension Cell Culture | |
| Surfactant | |
| No Phenol Red | |
| cGMP-compliant under the ISO 13485 standard | |
| Gibco | |
| Room Temperature |
| Mammalian Cells | |
| 10% | |
| Liquid | |
| Sterile-filtered | |
| Translational Research, Clinical Research | |
| 6 to 8 | |
| 100 mL | |
| Animal origin-free |
For Research Use Only. Not for use in diagnostic procedures.