missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Sf9 cells in Sf-900™ III SFM
Uused to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins
Supplier: Gibco™ 12659017
Alert:
Review the Material Safety Data Sheet before handling.
Description
Gibco Sf9 cells are adapted to serum-free suspension culture in Gibco Sf-900 III SFM.
- Recombinant protein expression from a variety of expression systems
- Good growth in adherent or suspension culture
- Small, regular size that generates even monolayers and plaques
- Documented lineage from a low passage Master Cell Bank
- Quality and performance testing
Baculovirus Production and Titering, Bioproduction, Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Sf9 and Sf21 Cell Culture, Viral Vaccine Production
Order Info
Shipping Condition: Dry Ice
Specifications
1 x 1.5 mL (1 x 107 cells/mL) Storage conditions: liquid nitrogen (vapor phase) Shipping conditions: frozen |
|
Insect Cells | |
Sf-900 III SFM (Serum-Free Media) | |
Insect Cells | |
Dry Ice |
Sf9 | |
S. frugiperda | |
Sf-900™ | |
1 Vial |
For Research Use Only. Not for use in diagnostic procedures.