missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Sf-900™ II SFM
Optimized for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS)
Supplier: Gibco™ 10902179
Description
Sf-900 II SFM is a serum-free, protein-free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells. This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines.
- Superior long-term, high-density growth
- Optimized for recombinant protein production
- Serum-free, protein-free, ready-to-use formulation
- Scalable in bioreactors
Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production
Specifications
Sf21, Sf9 | |
Protein-free, Serum-free | |
Insect Cell Serum Free Medium (SFM) | |
Glutamine | |
10 L | |
S. frugiperda, Spodoptera frugiperda |
Insect Cell | |
Liquid | |
Serum-free | |
Gibco™, Sf-900™, SynerGy™ | |
Room Temperature | |
Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.