Gibco™ Distilled Water DFS Item
High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents
Supplier:  Gibco™ 15230279
 View more versions of this product
Catalog No. 15230279
Explore available promotions
Description
Description
  • Suitable for use in cell culture and molecular biology applications: This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers
  • Prepared by distillation and filtration (0.1μm), with no added substances
  • Water quality monitored on a routine basis: Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly
  • Produced at a cGMP-compliant facility located in Paisley, Scotland, UK, registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard
Cell Culture, Mammalian Cell Culture
Specifications
Specifications
6 to 8
Universal Bag
Not DEPC-Treated
Membrane-Filtered
Gibco™
Cell Culture
Sterile-filtered
20 L
Room Temperature
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.