missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ TrypLE™ Express Enzyme (1X), no phenol red DFS Item

Unlike porcine trypsin, free of animal-derived components

Supplier:  Gibco™ 12604021

 View more versions of this product

Catalog No. 12604021


Explore available promotions

Description

Description

Cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts. Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells.

  • Gentle on cells
  • Room temperature-stable for 24 months
  • Easy to use
  • Animal origin-free
  • Gentle on cells, purity increases specificity due to the action of a single enzyme and reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts
  • Can be directly substituted for trypsin in existing protocols
  • Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS

Energy efficient, less hazardous, sustainable packaging

Cell Culture, Mammalian Cell Culture

Specifications

Specifications

EDTA
Mammalian Cells
1 X
Liquid
Sterile-filtered
Clinical Research, Translational Research
Check and Record
500 mL
Room Temperature
In vitro Bioassay
Storage conditions: 15°C to 30°C, protect from light
Alternate storage conditions: 2°C to 8°C or -5°C to -20°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Animal Origin-free
Adherent Cell Culture
Cell Culture Dissociation Reagent
No Phenol Red
Energy efficient, Sustainable packaging
Gibco™
Room Temperature Stable
Animal origin-free
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.