missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ TrypLE™ Express Enzyme (1X), phenol red
Animal origin-free, recombinant enzyme
Supplier: Gibco™ 12605010
Description
Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. TrypLE™ Express cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts.
- Gentle On Cells
- urity increases specificity due to the action of single enzyme, reducing damage caused by cleavage from enzymes present in some trypsin preparations and in other protease extracts
- Room-Temperature Stable for 24 months
- Ready to use
- Can be directly substituted for trypsin in existing protocols
- Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS
Energy efficient, less hazardous, sustainable packaging
Cell Culture, Mammalian Cell Culture
Specifications
EDTA | |
Mammalian Cells | |
1 X | |
Liquid | |
Sterile-filtered | |
Energy efficient, Sustainable packaging | |
Gibco™ | |
Room Temperature Stable | |
Animal origin-free |
Storage conditions: 15°C to 30°C, protect from light Alternate storage conditions: 2°C to 8°C or -5°C to -20°C Shipping conditions: Room temperature Shelf life: 24 months from date of manufacture |
|
Animal Origin-free | |
Adherent Cell Culture | |
Cell Culture Dissociation Reagent | |
Phenol Red, EDTA | |
Check and Record | |
100 mL | |
Room Temperature | |
In vitro Bioassay |
For Research Use Only. Not for use in diagnostic procedures.