missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Trypsin-EDTA (0.25%), phenol red DFS Item

Made from trypsin powder, irradiated mixture of proteases derived from porcine pancreas

$35.70 - $576.00

Specifications

Chelators EDTA
Content And Storage Storage conditions: -5°C to -20°C
Shipping conditions: Wet ice
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin
Form Liquid
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
25200056 DFS Item
View Documents
Gibco™
25200056
100 mL N/A
 
25200072 DFS Item
View Documents
Gibco™
25200072
500 mL N/A
 
25200114 DFS Item
View Documents
Gibco™
25200114
20 x 100 mL N/A
 
Description

Description

Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements.

  • Solutions tested for pH, osmolality, sterility, and performance
  • Raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis
  • Documented traceability available including lot traceability, animal origin certificates, lot analyses, irradiation certificates, viral inactivation summary, and supply chain transparency

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Compliance

Manufactured at a cGMP compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.

Specifications

Specifications

EDTA
Mammalian Cells
Liquid
Sterile-filtered
Phenol Red, EDTA
Gibco™
In vitro Bioassay
Storage conditions: -5°C to -20°C
Shipping conditions: Wet ice
Shelf life: 24 months from date of manufacture
Animal Origin
Cell Culture Dissociation Reagent
Sterile-filtered
270 - 320 mOsm/kg
Animal Origin
Videos
SDS
Documents

Documents

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.