missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Versene Solution
An EDTA solution for use as a gentle non-enzymatic cell dissociation reagent
Supplier: Gibco™ 15040066
Description
- Gibco Versene Solution 0.48 mM
- Formulated as 0.2 g EDTA(Na4) per liter of Phosphate Buffered Saline (PBS)
- Sustainable packaging
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Specifications
EDTA | |
Animal Origin-free | |
Sterile-filtered | |
cGMP for medical devices, 21 CFR Part 820 and ISO 13485 | |
Room Temperature |
Storage conditions: 2 to 8°C Shipping conditions: Ambient Shelf life: 18 months from date of manufacture |
|
Cell Culture Dissociation Reagent | |
EDTA | |
100 mL |
For Research Use Only. Not for use in diagnostic procedures.