missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Zeocin™ Selection Reagent DFS Item

Selection antibiotic, formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus

Supplier:  Gibco™ R25005

 View more versions of this product

Catalog No. R25005


Explore available promotions

Description

Description

  • Gibco™ Zeocin™ Selection Antibiotic is a formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus
  • Zeocin™ Selection Antibiotic acts by binding to and cleaving DNA, leading to cell death
  • It is used for selection of of mammalian, insect, yeast, bacterial, and plant cell cultures
  • Working concentration ranges between 50–400 μg/mL depending on the cell type
  • This product is supplied at 100 mg/mL in sterile water, in 8 x 1.25 mL vials
Specifications

Specifications

Storage conditions: -5 to -20°C
Shipping conditions: Ice
Mammalian Cell Culture, Insect Cell Culture
Antibiotic
Eukaryotic Selection/Stable Cell Line Generation, Bacterial Selection
50 mL
100 mg/mL
Liquid
Sterile-filtered
Zeocin™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.