Mopec™ Gigli Autopsy Saw Wire
Gigli Autopsy Saw Wire constructed of heavy duty steel and designed to be attached to Gigli saw handles.
Supplier:  Mopec™ AG023
Catalog No. 22-445-438
Explore available promotions
Description
Description
  • Made of standard grade stainless steel
  • For use with AG021 and AG121 Gigli saw handles
Specifications
Specifications
Stainless Steel
20 in.