Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Single™ Pipet Holder
Adhesive-backed hanger is designed for use where space is limited (fume hoods, sterile hoods, etc.)
Supplier:  Gilson™ F161406
Catalog No. F161406G
Explore available promotions
Description
Description
  • Sticks to any smooth, dry surface without damage
  • Resistant to chemicals and UV light
  • Holds any Gilson Pipette except the PIPETMAN™ Concept
Specifications
Specifications
White
Pipetteman™ complete pipetting system