Gilson™ EXPERT™ Universal Fit Pipette Tips
Featuring patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding when dispensing
Supplier:  Gilson™ F1733281
 View more versions of this product
Catalog No. F1733281G
Explore available promotions
Description
Description
  • Clear packaging allows easy identification of tips
  • Color-coded tip packaging for volume color matching to pipettes
  • Tips feature graduations at varying volumes allowing for quick-checks when aspirating samples
  • Economical and easy-to-use
  • Manufactured in sterile, clean-room environment
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Specifications
1 to 200 μL
8.255cm
Round
EXPERT
5μL
Sterile
0.57 mm
Yes
Polymer