Learn More
Ginkgolide B, Thermo Scientific Chemicals
Platelet-Activating Factor (PAF) antagonist
$190.95 - $644.23
Chemical Identifiers
CAS | 15291-77-7 |
---|---|
Molecular Formula | C20H24O10 |
Molecular Weight (g/mol) | 424.402 |
MDL Number | MFCD05662347 |
InChI Key | SQOJOAFXDQDRGF-WVXYHUDCSA-N |
Synonym | sqojoafxdqdrgf-maggbrlhsa-n |
PubChem CID | 132470872 |
SMILES | CC1C(=O)OC2C1(C34C(=O)OC5C3(C2O)C6(C(C5)C(C)(C)C)C(C(=O)OC6O4)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ60646MA
|
Thermo Scientific Chemicals
J60646MA |
10 mg |
N/A
|
|
|||||
AAJ60646MF
|
Thermo Scientific Chemicals
J60646MF |
50 mg |
N/A
|
|
|||||
Description
Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGinkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.
Solubility
Soluble in DMSO. Slightly soluble in water and ethanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.
Chemical Identifiers
15291-77-7 | |
424.402 | |
SQOJOAFXDQDRGF-WVXYHUDCSA-N | |
132470872 |
C20H24O10 | |
MFCD05662347 | |
sqojoafxdqdrgf-maggbrlhsa-n | |
CC1C(=O)OC2C1(C34C(=O)OC5C3(C2O)C6(C(C5)C(C)(C)C)C(C(=O)OC6O4)O)O |
Specifications
15291-77-7 | |
C20H24O10 | |
10 mg | |
sqojoafxdqdrgf-maggbrlhsa-n | |
SQOJOAFXDQDRGF-WVXYHUDCSA-N | |
424.402 | |
424.4 | |
Ginkgolide B |
White | |
MFCD05662347 | |
14,4426 | |
Soluble in DMSO; Slightly soluble in water and ethanol | |
CC1C(=O)OC2C1(C34C(=O)OC5C3(C2O)C6(C(C5)C(C)(C)C)C(C(=O)OC6O4)O)O | |
132470872 | |
Powder |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only