missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ GL45 Polypropylene Screw Caps

Linerless plug seal design for efficient sealing

$167.72 - $242.58

Specifications

Material Polypropylene
Sterility Non-sterile
Thread Size (Metric) GL 45
For Use With (Equipment) Storage bottle
Closure Material Polypropylene
View More Specs

Products 4
Catalog Number Mfr. No. Color For Use With (Equipment) No. per Case Price Quantity  
Catalog Number Mfr. No. Color For Use With (Equipment) No. per Case Price Quantity  
06-414-3B
View Documents
Corning™
139545LTC2
Light Gray Storage bottle 20/Cs.
Case of 20 Each for $234.21
 
06-414-4
View Documents
Corning™
139545LTMC
Gray Storage bottle 10/Cs.
Case of 10 Each for $167.72
 
06-414-3C
View Documents
Corning™
139545LTC3
Green Storage bottle 20/Cs.
Case of 20 Each for $231.94
 
06-414-3A
View Documents
Corning™
139545LTC1
Purple Storage bottle 20/Cs.
Case of 20 Each for $242.58
 
Description

Description

These are various sizes and types of screw caps for use with storage bottles.

Specifications

Specifications

Polypropylene
GL 45
Polypropylene
Non-sterile
Storage bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'