missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Glass Beads

Contains replacement reagents to run all Saftest™ test kits.

Supplier:  MP Biomedicals™ 07DP1070

Catalog No. MP07DP1070


Explore available promotions

Description

Description

  • For use with SafTest analyzer
  • Used with all six SafTest test kits
  • Appropriate for all testing endpoints
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Application area: Oil/fat degradation
Specifications

Specifications

Glass
One tube (5mm sized beads)
Videos
SDS
Documents

Documents

Promotions

Promotions