missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Concave Mirror

Round, optically worked Glass Concave Mirror with a silvered back and protective coating. Designed for physical science demonstrations and experiments.

$7.77 - $77.63

Specifications

Material Glass
For Use With (Application) Physical Science Demonstrations and Experiments
Product Type Mirror
View More Specs

Products 15
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Focal Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Focal Price Quantity  
S27362
View Documents
Eisco™
PH0520A
50 mm 50 mm
Each for $13.27
Only null left
 
S27686
View Documents
Eisco™
PH0520C
50 mm 100 mm
Each for $10.67
Only null left
 
S27687
View Documents
Eisco™
PH0520D
50 mm 150 mm
Each for $10.67
Only null left
 
S27363
View Documents
Eisco™
PH0520B
50 mm 75 mm
Each for $7.77
Only null left
 
S27688
View Documents
Eisco™
PH0520E
50 mm 200 mm
Each for $10.67
Only null left
 
S27689
View Documents
Eisco™
PH0520F
50 mm 300 mm
Each for $10.67
Only null left
 
S27690
View Documents
Eisco™
PH0520H
75 mm 75 mm
Each for $20.40
Only null left
 
S27365
View Documents
Eisco™
PH0520I
75 mm 100 mm
Each for $15.37
Only null left
 
S27366
View Documents
Eisco™
PH0520J
75 mm 150 mm
Each for $15.37
Only null left
 
S27691
View Documents
Eisco™
PH0520K
75 mm 200 mm
Each for $20.27
Only null left
 
S27692
View Documents
Eisco™
PH0520L
75 mm 300 mm
Each for $20.27
Only null left
 
S27693
View Documents
Eisco™
PH0520M
100 mm 100 mm
Each for $31.77
Only null left
 
S27694
View Documents
Eisco™
PH0520N
100 mm 150 mm
Each for $31.77
Only null left
 
S27695
View Documents
Eisco™
PH0520O
150 mm 150 mm
Each for $77.63
Only null left
 
S27696
View Documents
Eisco™
PH0520P
150 mm 300 mm
Each for $77.63
Only null left
 
Description

Description

  • For educational use only
Specifications

Specifications

Glass
Physical Science Demonstrations and Experiments
Mirror
Videos
SDS
Documents

Documents