Glass Funnel with Vent
Heavy molded-glass funnel with vent.
Supplier:  Customer Channels Group (Ccg) S34580
 View more versions of this product
Catalog No. S34580
Explore available promotions
Description
Description
  • Give displaced air a way to escape from the receiving container so liquid can flow in smoothly
  • Made with smooth rims and inside rib
  • The air vent on the stem makes pouring easy
Specifications
Specifications
11.5 cm
125 mL
Glass
Gives displaced air a way to escape from the receiving container so liquid can flow in smoothly