missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GSC Go Science Crazy Glass Coverslips DFS Item

Pre-cleaned coverslips.

Supplier:  GSC Go Science Crazy 413051

 View more versions of this product

Catalog No. S17466A


Explore available promotions

Description

Description

  • Plain glass
  • Good quality
  • Coverslip thickness: 0.13 to 0.17mm (size No. 1)
  • Coverslip sizes: 18 x 18mm (S13604) or 22 x 22mm (S175211A) 
Specifications

Specifications

Glass
25 mm
75 x 25 mm
75 mm
1 to 1.2 mm
Videos
SDS
Documents

Documents

Promotions

Promotions