MilliporeSigma™ Supelco™ Glass Packed GC Column (fits Agilent, Configuration A), Phase: 10 Carbowax 20M, Matrix 100/120 SUPELCOPORT Support
Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness.
Supplier: MilliporeSigma™ Supelco™ 12478U
Description
This column fits Agilent 5880, 5890, 6890. 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).Specifications
Packed GC | |
Gas Chromatography (GC) | |
Glass Column | |
2 mm |
0.25 in. | |
2 m | |
Carbowax 20 M, Matrix 100/120 SUPELCOPORT Support |