MilliporeSigma™ Supelco™ Glass Packed GC Column (fits Agilent, Configuration A), Phase: 4 Carbowax 20M + 0.8 KOH, Matrix 60/80 Carbopack B Support
Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness.
Supplier:  MilliporeSigma™ Supelco™ 26030U
Catalog No. 111005411
Explore available promotions
Description
Description
This column fits Agilent 5880, 5890, 6890. 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).
Specifications
Specifications
Packed GC
Gas Chromatography (GC)
Glass Column
2 mm
0.25 in.
1.8 m
Carbowax 20 M + 0.8 KOH, Matrix 60/80 Carbopack B Support