Eisco™ Glass Pear Shaped Separating Funnel with Glass Key Stopcock, Interchangable Plastic Stopper
Pear shaped separatory funnels made from high quality borosilicate glass. Designed with integrated glass key stopcock and interchangable plastic stopper. Autoclavable.
$243.24 - $243.24
Specifications
Material | Borosilicate Glass |
---|---|
Autoclavable | Autoclavable |
Color | Clear |
Graduated | No |
Includes | Stopper |
Includes: Glass Funnel, Glass Stopcock, Interchangable Plastic Stopper
Catalog Number | Mfr. No. | Capacity (Metric) | Joint Number | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Joint Number | Price | Quantity | ||||
S41694
|
Eisco™
CH0482F |
2000 mL | 29/32 |
Each for $243.24
|
|
||||
Description
- Separatory funnel made from high quality borosilicate glass
- Pear shaped
- Includes integrated glass key stopcock
- Plastic interchangeable stopper for top
- Autoclavable
Warranty
1 year limited manufacturer warranty
Specifications
Borosilicate Glass | |
Clear | |
Stopper |
Autoclavable | |
No | |
1 |