missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Glass Reagent Bottles with Polypropylene Stopper
Clear glass reagent bottles with polypropylene stoppers are ideal for storing liquid solutions in the laboratory. The polypropylene stoppers are acid proof. Both the glass bottles and stoppers are autoclavable.
$15.65 - $125.03
Specifications
Color | Clear |
---|---|
For Use With (Application) | Solution Preparation, Solution Storage |
Includes | Clear glass bottle, polypropylene stopper |
Material | Borosilicate Glass |
Quantity | 1 |
Includes: Clear glass bottle, polypropylene stopper
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
S39281
|
Eisco™
CH0160A |
30 mL |
Each for $15.65
|
|
|||||
S39282
|
Eisco™
CH0160B |
60 mL |
Each for $17.56
|
|
|||||
S39283
|
Eisco™
CH0160C |
125 mL |
Each for $26.26
|
|
|||||
S39285
|
Eisco™
CH0160E |
500 mL |
Each for $40.06
|
|
|||||
S39287
|
Eisco™
CH0160G |
2 L |
Each for $125.03
|
|
|||||
Description
- Made of high quality borosilicate 3.3 glass
- Narrow mouth with acid proof polypropylene stopper
- Bottles and stoppers are autoclavable
Warranty
1 year limited manufacturer warranty
Specifications
Clear | |
Clear glass bottle, polypropylene stopper | |
1 | |
Yes | |
Reusable | |
Narrow | |
Reagent Bottle | |
Unlabeled | |
Stopper |
Solution Preparation, Solution Storage | |
Borosilicate Glass | |
Non-sterile | |
Autoclavable | |
No | |
Cylindrical | |
Liquids | |
Polypropylene |