missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Glass Reagent Bottles with Polypropylene Stopper
Clear glass reagent bottles with polypropylene stoppers are ideal for storing liquid solutions in the laboratory. The polypropylene stoppers are acid proof. Both the glass bottles and stoppers are autoclavable.
Supplier: Eisco™ CH0160E
Includes: Clear glass bottle, polypropylene stopper
Description
- Made of high quality borosilicate 3.3 glass
- Narrow mouth with acid proof polypropylene stopper
- Bottles and stoppers are autoclavable
Warranty
1 year limited manufacturer warranty
Specifications
500 mL | |
16.9 oz. | |
3.4 in. | |
Clear glass bottle, polypropylene stopper | |
86.4mm | |
Non-sterile | |
86.4mm | |
Autoclavable | |
No | |
Cylindrical | |
Reagent Bottle | |
6 in. | |
Polypropylene | |
86.4 mm |
500 mL | |
Clear | |
Solution Preparation, Solution Storage | |
Borosilicate Glass | |
1 | |
226.79 g | |
Yes | |
Reusable | |
Narrow | |
155 mm | |
Liquids | |
Unlabeled | |
24/29 mm | |
Stopper |