missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Glass Reagent Bottles with Polypropylene Stopper

Clear glass reagent bottles with polypropylene stoppers are ideal for storing liquid solutions in the laboratory. The polypropylene stoppers are acid proof. Both the glass bottles and stoppers are autoclavable.

Supplier:  Eisco™ CH0160G

 View more versions of this product

Catalog No. S39287


Explore available promotions

Includes: Clear glass bottle, polypropylene stopper

Description

Description

  • Made of high quality borosilicate 3.3 glass
  • Narrow mouth with acid proof polypropylene stopper
  • Bottles and stoppers are autoclavable

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

2 L
67.6 oz.
5.4 in.
Clear glass bottle, polypropylene stopper
137.2mm
Non-sterile
Yes
Reusable
Narrow
259.1 mm
Liquids
Unlabeled
34/35 mm
Stopper
2 L
Clear
Solution Preparation, Solution Storage
Borosilicate Glass
1
137.2mm
Autoclavable
No
Cylindrical
Reagent Bottle
10.2 in.
Polypropylene
137.2 mm
SDS
Documents

Documents

Promotions

Promotions