Glass Vials, Inc. Glass Sample Vial with TFE-Lined Cap
Flint glass with positive sealing, TFE-lined screw cap
Supplier:  Glass Vials, Inc. V1235CTFE
Catalog No. 03-339
Explore available promotions
Description
Description
  • 1/2 dr. (1.8mL) capacity
  • GPI Thread Finish 8-425
  • Autoclavable
1/2 O.D. × 1 3/8 in. high (12 × 35mm)
  • Preparation and storage of tester strains for determination of carcinogenic compounds via Ames test
  • Toxicology studies
Specifications
Specifications
Silica Glass
0.5 dr.
Screw Cap
Product Suggestions
Product Suggestions