MilliporeSigma™ Supelco™ Glass Wool
Often used in GC inlet liners to improve sample vaporization, and/or to keep non-volatile material from entering the column
$86.71 - $1170.30
Specifications
For Use With (Application) Packed GC Columns, Solvent Desorption Tubes, Thermal Desorption Tubes, and Purge Traps to Retain Adsorbent Beds
Material Glass Fiber
View More Specs
Products 5
Catalog Number Mfr. No. Product Type Weight (Metric) Price Quantity  
Catalog Number Mfr. No. Product Type Weight (Metric) Price Quantity  
111020909
View Documents
MilliporeSigma™ Supelco™
SGMA20409
GC Accessories and Adsorbents 10 g
Each for $113.63
111020928
View Documents
MilliporeSigma™ Supelco™
SGMA20384
Non-Treated 50 g
Each for $86.71
111008042
View Documents
MilliporeSigma™ Supelco™
21688U
GC Accessories and Adsorbents 100 g
Each for $1,170.30
111020914
View Documents
MilliporeSigma™ Supelco™
SGMA20383
Phosphoric Acid Treated 50 g
Each for $107.36
111020786
View Documents
MilliporeSigma™ Supelco™
SGMA20410
Silanized 250 g
Each for $487.92
Description
Description
  • Made up of short glass fibres
  • Non-flammable and non-carcinogenic
  • Widely considered as a thermal insulation material in ships, automobiles, air conditioning ducts, and water supply pipes
  • Suitable as noise absorbing material in concert halls, studios, and movie theatres
  • Can be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain adsorbent beds
Specifications
Specifications
Packed GC Columns, Solvent Desorption Tubes, Thermal Desorption Tubes, and Purge Traps to Retain Adsorbent Beds
Glass Fiber
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'