Brady™ GlobalMark™ Industrial Label Maker Tapes
Designed with maximum durability for harsh, industrial environments. Brady GlobalMark Industrial Label Maker Tapes is made of B-595 vinyl and adheres to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics.
Supplier:  Brady™ 113161
 View more versions of this product
Catalog No. 19-107-986
Explore available promotions
Description
Description
  • Made of B-595 vinyl with permanent adhesive
  • With preprinted strips
  • Applied to textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes
Specifications
Specifications
Compatible Surface Types: Powder coated, smooth, textured, rough, irregular, curved
Red, White
Cartridge
Industrial label maker tape
100 ft. x 4 in. with 1 in. stripe
8 to 10 Years
30.4 m
10.16 cm
Acrylic, Permanent, Cold-temperature
Gloss
Vinyl Film (B-595)
GlobalMark Label Maker
0.004 in.
100 ft.
4 in.