Globe Scientific Accessories for Spectrophotometer Cuvettes
For use with Spectophotometer cuvettes
Supplier:  Globe Scientific 111167
 View more versions of this product
Catalog No. 21-200-260
Explore available promotions
Description
Description
  • Polyethylene made 12-place cuvette holder of 170 x 50 x 15mm dimensions
  • LDPE snap caps for square cuvettes for added protection against spillage and contamination
  • Polystyrene disposable 90mm stirring stick for access to the bottom of cuvettes
Specifications
Specifications
Plug Cap
Spectrophotometer cuvettes
LDPE
10 mm