Globe Scientific Capillary Tube Accessories
For use with capillary tubes
Supplier:  Globe Scientific 51674
Catalog No. 22-170-406
Explore available promotions
Description
Description
For use with capillary tubes
  • Pipet bulb helps to expel blood from capillary tubes
  • Used for drawing blood directly from 13mm or 16mm blood collection tube
Specifications
Specifications
Bulb
Capillary Tubes