Globe Scientific EZ-Rate™ Polystyrene ESR Pipet
For Westergren ESR determinations directly in 13mm sodium citrate blood collection tubes
Supplier:  Globe Scientific 3479
Catalog No. 21-200-212
Explore available promotions
Description
Description
  • Polystyrene made, gaduated
  • With self sealing, fibrous plug located at zero mark
  • For 100 tests
Specifications
Specifications
EZ-Rate™ ESR Westergren Pipet
For Westergren ESR determinations directly in 13mm Sodium Citrate Blood collection tubes
Self sealing fibrous plug
100 U
Product Suggestions
Product Suggestions