missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Globe Scientific Polyethylene 16mm Plug Stoppers
Provide a leak-resistant seal
Supplier: Globe Scientific 116142Y
Description
- Made of polyethylene
- Assorted colors available
Specifications
Plug Stopper Cap | |
Yellow | |
16 mm |
Polyethylene | |
1000 |