Globe Scientific Sterile Rigid Inoculating Loops with Needles
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media. Molded from a special medical grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Supplier: Globe Scientific 2865F
Description
- Certificate of Calibration comes with each case
- Gamma Sterilized
Specifications
Single-use | |
Polystyrene | |
Yellow | |
Needle and calibration certificate | |
Peel Pack |
Sterile | |
10 μL | |
Inoculating Loop, Rigid | |
10 |