Globe Scientific Sterile Rigid Inoculating Loops with Needles
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media. Molded from a special medical grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Supplier:  Globe Scientific 2851F
 View more versions of this product
Catalog No. 22-170-209
Explore available promotions
Description
Description
  • Certificate of Calibration comes with each case
  • Gamma Sterilized
Specifications
Specifications
Single-use
Polystyrene
Blue
Needle and calibration certificate
Individually Wrapped
Sterile
1 μL
Inoculating Loop, Rigid
Each