Globe Scientific Universal Snap Caps
Provides a leak-resistant closure
Supplier:  Globe Scientific 118115R
 View more versions of this product
Catalog No. 22-171-661
Explore available promotions
Description
Description
  • Precision molded low-density polyethylene made
  • Fits most 12, 13 and 16mm plastic test tubes
  • Avoids cross-contamination, protects lab work areas against infection and spillage
  • Not autoclavable
  • Assorted colors available for sample identification
Specifications
Specifications
Snap Cap
Polyethylene
Universal
Not Autoclavable
Red
Recapping 12, 13 and 16 mm evacuated glass blood collection test tubes