missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega GloSensor™ cAMP Assay DFS Item

The GloSensor cAMP Assay is based on a genetically modified form of firefly luciferase into which a cAMP-binding protein moiety has been inserted. This live-cell assay excels at kinetic and modulation studies of signaling through cAMP.

$797.15 - $3217.57

Specifications

Detection Method Luminescence
For Use With (Application) Full and partial agonists and antagonists as well as inverse agonists and allosteric modulators
Form Fluid
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Product Type Price Quantity  
Catalog Number Mfr. No. Quantity Product Type Price Quantity  
PRE1291 DFS Item
View Documents
Promega
E1291
250 mg GloSensor cAMP Reagent
Each for $3,217.57
 
PRE1290 DFS Item
View Documents
Promega
E1290
25 mg GloSensor cAMP Reagent
Each for $797.15
 
PRE2301 DFS Item
View Documents
Promega
E2301
20 μg pGlo Sensor 22F cAMP Plasmid
Each for $1,233.28
 
PRE1171 DFS Item
View Documents
Promega
E1171
20 μg pGlo Sensor 20F cAMP plasmid
Each for $1,253.04
 
Description

Description

  • Live-Cell Biosensors for cAMP, cGMP or Protease Activity
  • Biosensors enable more complete analysis of receptor biology
  • Simple, scalable assay protocol
  • High Z´ and large signal:background ratio
Specifications

Specifications

Luminescence
Full and partial agonists and antagonists as well as inverse agonists and allosteric modulators
Fluid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

  • By use of this product, researcher agrees to be bound by the terms of this limited use statement. If the researcher is not willing to accept the conditions of this limited use statement, and the product is unused, Promega will accept return of the unused product and provide the researcher with a full refund.
  • Not for use in live animals—For use in live animals contact Promega regarding the availability of licenses. Researchers may use this product and its derivatives in their own research solely for the luminescent determination of cAMP levels in vitro or within cells (excluding uses in living animals). Researchers shall have no right to modify or otherwise create variations of the nucleotide sequence encoding the GloSensor cAMP Reporter of this product (i.e., the bioluminescent chimeric protein comprising luciferase and cAMP-binding domains). Researchers may transfer derivatives to others provided that recipients agree to be bound by the conditions of this limited use statement prior to such transfer. No other use or transfer of this product or derivatives is authorized without the prior express written consent of Promega. In addition, researchers must either: (1) use the GloSensor cAMP Reagent purchased from Promega for all determinations of luminescence activity of this product and derivatives; or (2) contact Promega to obtain a license for use of the product and derivatives in conjunction with luminescent assay reagents not purchased from Promega. Promega makes no representations or warranties of any kind, either expressed to implied, including for merchantability or fitness for a particular purpose with regards to the product. The terms of this agreement shall be governed under the laws of the State of Wisconsin, USA.
  • Mfr. No. E2301, E1171: U.S. Patent 5,670,356.
  • Mfr. No. E2301, E1171: Australian Patent 2001 285278 and other patents pending.
  • Mfr. No. E1261: Commercial use of this cell line requires a license from AdVec Inc.
  • Mfr. No. E1261: HEK293 cells were obtained under license from AdVec Inc.
  • Mfr. Nos. E1261, E2301, E1171: The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.