missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Poltex Glove Box Holder ST
Holders are constructed of break-resistant PETG and can be mounted horizontally or vertically. Each glove box has its own individual compartment for easy replacement of empty boxes.
Supplier: Poltex 2GBSTM
Description
- Constructed of clear, break-resistant PETG
- Each box has its own individual compartment for easy replacement of empty boxes
- Choose from either a magnet or keyhole mount (hardware not included)
- Holder can be positioned vertically or horizontally
- Glove boxes are shown for demonstration purposes only
Specifications
Glove Box Holder | |
9 x 3.95 x 11 in. | |
22.9 cm | |
10 cm | |
27.9 cm | |
PETG | |
2 |
Clear | |
9 in. | |
3.95 in. | |
11 in. | |
Disposable Glove Boxes | |
Magnetic Mount | |
2 Boxes |