missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poltex Glove Box Holder ST DFS Item

Holders are constructed of break-resistant PETG and can be mounted horizontally or vertically. Each glove box has its own individual compartment for easy replacement of empty boxes.

Supplier:  Poltex 4GBSTW

 View more versions of this product

Catalog No. 22000142


Explore available promotions

Description

Description

  • Constructed of clear, break-resistant PETG
  • Each box has its own individual compartment for easy replacement of empty boxes
  • Choose from either a magnet or keyhole mount (hardware not included)
  • Holder can be positioned vertically or horizontally
  • Glove boxes are shown for demonstration purposes only
Specifications

Specifications

Glove Box Holder
9 x 3.95 x 22 in.
22.9 cm
10 cm
55.9 cm
PETG
4
Clear
9 in.
3.95 in.
22 in.
Disposable Glove Boxes
Keyhole Slots
4 Boxes
Videos
SDS
Documents

Documents

Promotions

Promotions