Invitrogen™ GLP-1 Multispecies ELISA Kit DFS Item
Supplier:  Invitrogen™ BMS2194DUP
Catalog No. BMS2194
Explore available promotions
Description
Description
GCG is also known as GLP1, or Glucagon. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis.It is mapped to 2q36-2q37. GLP1, also known as 7-37 for the codons of the preproglucagon molecule which encode it, renders pancreatic beta-cells 'glucose-competent' and may be useful in the treatment of noninsulin-dependent diabetes mellitus. Also, GLP1 is a potent insulin secretagogue.It plays a major role in the enteroinsular axis, accounting, for example, for the finding that plasma insulin levels accompanying oral intake of glucose are greater than those observed when glucose is given intravenously.
Specifications
Specifications
0.8 to 50.0 pg/mL
Biotin
Plasma, Serum, Supernatant
Gcg; Glicentin; Glicentin-related polypeptide; GLP1; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP1/2; GLP2; GLP-2; Glu; Glucagon; Glucagon like peptide 1; glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; Incretin hormone; MGC138331; OXM; OXY; Oxyntomodulin; PPG; preproglucagon
GCG
3.8%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
0.32 pg/mL
ELISA
Colorimetric Microplate Reader
P01275
3.8%
Pre-coated 96 well plate
Standard
Sample Diluent
Assay Buffer concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers
96 tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
GLP-1
3 hr 30 min