missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gluconic Acid, 50 wt. % Solution in Water, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 119890010

 View more versions of this product

Catalog No. AC119890010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gluconic acid
White to Yellow
MFCD00004240
15, 4492
RGHNJXZEOKUKBD-SQOUGZDYSA-N
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
10690
196.16
1.23g/mL
Glass bottle
1.22 to 1.25 (20°C)
526-95-4,7732-18-5
C6H12O7
03, 542
gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate
C(C(C(C(C(C(=O)O)O)O)O)O)O
196.155
CHEBI:33198
49 to 55% (Titrimetry other)
1 L
Crystals or Crystalline Powder

Chemical Identifiers

526-95-4
196.155
RGHNJXZEOKUKBD-SQOUGZDYSA-N
10690
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
C6H12O7
MFCD00004240
gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate
CHEBI:33198
C(C(C(C(C(C(=O)O)O)O)O)O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 208-401-4

RTECSNumber : LZ5057100

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only