missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gluconic acid, potassium salt, 99%, Thermo Scientific Chemicals

$96.05 - $96.05

Chemical Identifiers

Molecular Formula C6H11KO7
Molecular Weight (g/mol) 234.25
MDL Number MFCD00064211
InChI Key HLCFGWHYROZGBI-JJKGCWMISA-M
Synonym potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal
PubChem CID 16760467
IUPAC Name potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229322500
SDS View Documents
Thermo Scientific Chemicals
229322500
250 g Plastic bottle
Each for $96.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

C6H11KO7
MFCD00064211
potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal
potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
234.25
HLCFGWHYROZGBI-JJKGCWMISA-M
16760467
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+]

Specifications

178°C to 181°C
3% max. (105°C, 4 hrs) (vacuum)
Plastic bottle
HOCH2[CH(OH)]4CO2K
250 g
+14.10° (21°C c=1.2,H2O)
potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal
HLCFGWHYROZGBI-JJKGCWMISA-M
potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
16760467
0.99
Gluconic acid, potassium salt, 0.99
White
Authentic
C6H11KO7
MFCD00064211
15, 4492
+ 14.1
Solubility in water: soluble in water. Other solubilities: practically insoluble in abs. alcohol,ether,,benzene and chloroform
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+]
234.25
234.25
Crystalline or Granular Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-074-2

RTECSNumber : LZ5230000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only