Glucose-Glutamic Acid, For BOD (Sterilized), Certified, 198.0mg/L BOD ±30.5mg/L, LabChem™
Supplier:  LabChem LC14845246
 View more versions of this product
Catalog No. LC14845246
Explore available promotions
Description
Description
Specifications
Specifications
Glucose-Glutamic Acid
For BOD, Sterilized
50-99-7,56-86-0
198.0mg/L BOD ± 30.5mg/L
Soluble in water
Certified
Vial
24 x 6 mL
Colorless
99.97,0.015,0.015
C6H12O6
GGA
180.15
Passes Test
Nitrogen oxides; Carbon monoxide; Carbon dioxide
Liquid