missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glucose-Glutamic Acid, For BOD (Sterilized), Certified, 198.0mg/L BOD ±30.5mg/L, LabChem™
Supplier: LabChem LC14845486
Specifications
Glucose-Glutamic Acid | |
For BOD, Sterilized | |
50-99-7,56-86-0 | |
198.0mg/L BOD ± 30.5mg/L | |
Soluble in water | |
Certified | |
Vial | |
48 x 6 mL |
Colorless | |
99.97,0.015,0.015 | |
C6H12O6 | |
GGA | |
180.15 | |
Passes Test | |
Nitrogen oxides; Carbon monoxide; Carbon dioxide | |
Liquid |