Glucose-Glutamic Acid Solution, Standard Check Solution for Biochemical Oxygen Demand (BOD), Ricca Chemical
Supplier:  Ricca Chemical Company 3255246
 View more versions of this product
Catalog No. 3255246
Explore available promotions
Description
Description
Specifications
Specifications
Glucose-Glutamic Acid Solution
50-99-7,56-86-0,7732-18-5
24 x 6 mL
100.0,0.02,0.02
C6H12O6
C(C(C(C(C(C=O)O)O)O)O)O
180.156
CHEBI:42758
Standard Check Solution for Biochemical Oxygen Demand (BOD)
Glass Vial
98.0,0.02,0.01
Colorless
GZCGUPFRVQAUEE-SLPGGIOYSA-N
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
107526
Liquid
Chemical Identifiers
50-99-7
180.156
107526
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
C6H12O6
GZCGUPFRVQAUEE-SLPGGIOYSA-N
CHEBI:42758
C(C(C(C(C(C=O)O)O)O)O)O
Product Suggestions
Product Suggestions
Videos
SDS Documents