Learn More
Glufosinate ammonium, 95%, Thermo Scientific Chemicals
Glufosinate ammonium, 95%, CAS # 77182-82-2, also known as glufosinate, is a non-selective phosphorous herbicide.
Supplier: Thermo Scientific Chemicals J66186MD
Description
Used as broad spectrum post-emergence agrochemical for grapes, orchards, plantations, ornamentals, and non-cropland; Also used as crop desiccant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Glufosinate ammonium is a compound which exhibits broad-spectrum herbicidal properties through inhibition of glutamine synthetase, which disrupts cell membranes
Application
• Glufosinate ammonium acts as a non-selective herbicide
Specifications
Glufosinate Ammonium | |
Powder | |
215°C | |
250 mg | |
C5H15N2O4P | |
519°C | |
14,7339 | |
Soluble in water | |
[NH4+].CP(O)(=O)CCC(N)C([O-])=O | |
198.16 | |
198.16 |
Glufosinate ammonium | |
77182-82-2 | |
White | |
>110°C (230°F) | |
MFCD00055562,MFCD00211362 | |
8163399 | |
DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt | |
ZBMRKNMTMPPMMK-UHFFFAOYNA-N | |
ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate | |
122130249 | |
95% |
Chemical Identifiers
77182-82-2 | |
198.16 | |
ZBMRKNMTMPPMMK-UHFFFAOYNA-N | |
122130249 | |
[NH4+].CP(O)(=O)CCC(N)C([O-])=O |
C5H15N2O4P | |
MFCD00055562,MFCD00211362 | |
DL-Phosphinothricin; 2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt | |
ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate |
Safety and Handling
GHS H Statement
H360-H373-H302-H312-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H360-H373
EINECSNumber : 278-636-5
RTECSNumber : EK7713600
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only