missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glutaconaldehydedianil Hydrochloride 98.0+%, TCI America™
Supplier: TCI America G0201250G
Specifications
Glutaconaldehydedianil Hydrochloride | |
140°C | |
C17H17ClN2 | |
250 g | |
VUCMMJBDNXZQDJ-ZUJIUJENSA-N | |
hydrogen N-[(1E,3E)-5-(phenylimino)penta-1,3-dien-1-yl]aniline chloride | |
5702667 | |
≥98.0% (N) |
1497-49-0 | |
Purple-Red | |
MFCD00012630 | |
hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride | |
[H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1 | |
284.79 | |
284.79 | |
Crystalline Powder |
Chemical Identifiers
1497-49-0 | |
284.79 | |
VUCMMJBDNXZQDJ-ZUJIUJENSA-N | |
5702667 | |
[H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1 |
C17H17ClN2 | |
MFCD00012630 | |
hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride | |
hydrogen N-[(1E,3E)-5-(phenylimino)penta-1,3-dien-1-yl]aniline chloride |
Safety and Handling
TSCA : Yes