missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaraldehyde, 25% aq. soln., Thermo Scientific Chemicals

$55.73 - $241.64

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA17876AE
View Documents
Thermo Scientific Chemicals
A17876AE
100 mL
Each for $55.73
 
AAA17876AP
View Documents
Thermo Scientific Chemicals
A17876AP
500 mL
Each for $101.37
 
AAA178760F
View Documents
Thermo Scientific Chemicals
A178760F
2500 mL
Each for $241.64
 
Description

Description

Glutaraldehyde is used for cross-linking proteins and polyhydroxy materials. Also used as a fixative for tissues and as an amine-reactive homobifunctional crosslinker. It is also used in coupling reaction of biotin hydrazides to nucleic acids which is useful for conjugating fluorescent hydrazides and hydroxylamines to DNA and for stabilization ofproteins on agarose beads,activation of polystyrene and glass, for immobilization of antibodies and antigens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaraldehyde is used for cross-linking proteins and polyhydroxy materials. Also used as a fixative for tissues and as an amine-reactive homobifunctional crosslinker and as a disinfectant for medical equipment. It is also used in coupling reaction of biotin hydrazides to nucleic acids which is useful for conjugating fluorescent hydrazides and hydroxylamines to DNA and for stabilization ofproteins on agarose beads,activation of polystyrene and glass, for immobilization of antibodies and antigens. It is a disinfectant, which is rapidly effective against vegetative forms of Gram-positiveand Gram-negative bacteria.

Solubility
Miscible with water, ethanol, benzene, ether, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.

Notes
Heat and air sensitive. Incompatible with strong acids, strong bases and strong oxidizing agents.
Specifications

Chemical Identifiers

111-30-8
100.12
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
3485
pentanedial
C5H8O2
MFCD00007025
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
CHEBI:64276
O=CCCCC=O

Specifications

111-30-8
0.25
1.373
MFCD00007025
UN2922
14,4472
Miscible with water,ethanol,benzene,ether,acetone,dichloromethane,ethylacetate,isopropanol,n-hexane and toluene.
O=CCCCC=O
100.12
CHEBI:64276
25% aq. soln.
1.062
C5H8O2
OHC(CH2)3CHO
100 mL
605390
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
pentanedial
3485
100.12
Glutaraldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H314-H302-H317

P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H332-H314-H317-H334

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 203-856-5

RTECSNumber : MA2450000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only