Learn More
Glutaraldehyde, 50% aq. soln., Thermo Scientific Chemicals
$57.55 - $376.88
Chemical Identifiers
CAS | 111-30-8 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00007025 |
InChI Key | SXRSQZLOMIGNAQ-UHFFFAOYSA-N |
Synonym | glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde |
PubChem CID | 3485 |
ChEBI | CHEBI:64276 |
IUPAC Name | pentanedial |
SMILES | O=CCCCC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1050022
|
Thermo Scientific Chemicals
A1050022 |
100 g |
Each for $57.55
|
|
|||||
AAA1050036
|
Thermo Scientific Chemicals
A1050036 |
500 g |
Each for $98.25
|
|
|||||
AAA105000E
|
Thermo Scientific Chemicals
A105000E |
2500 g |
Each for $376.88
|
|
|||||
Description
Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.
Solubility
Miscible with water, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.
Notes
Incompatible with strong oxidisng agents, strong acids and strong bases.
Chemical Identifiers
111-30-8 | |
100.12 | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
3485 | |
pentanedial |
C5H8O2 | |
MFCD00007025 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
CHEBI:64276 | |
O=CCCCC=O |
Specifications
111-30-8 | |
1.123 g/mL | |
C5H8O2 | |
MFCD00007025 | |
605390 | |
Miscible with water,acetone,dichloromethane,ethylacetate,isopropanol,n-hexane and toluene. | |
O=CCCCC=O | |
100.12 | |
CHEBI:64276 | |
Glutaraldehyde |
∽-21°C | |
100°C | |
1.421 | |
100 g | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
100.12 |
Safety and Handling
P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H301-H314-H317-H330-H334
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 203-856-5
RTECSNumber : MA2450000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only