missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaraldehyde Solution, 25% (Certified), Fisher Chemical™

$753.00 - $2234.17

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
O2957-1
SDS View Documents
Thermo Fisher Scientific
O29571
1 L Glass Bottle
Each for $753.00
Case of 6 Each for $4,518.00
 
O2957-4
SDS Accessories View Documents
Thermo Fisher Scientific
O29574
4 L Glass Bottle
Each for $2,234.17
Case of 4 Each for $8,936.68
 
Description

Description

Specifications

Chemical Identifiers

111-30-8
100.12
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
3485
pentanedial
C5H8O2
MFCD00007025
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
CHEBI:64276
O=CCCCC=O

Specifications

Liquid
100°C
1.060g/cm³
16.4mmHg at 20°C
24.0 to 26.5 %
HC(O)(CH2)3CHO
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
O=CCCCC=O
100.12
CHEBI:64276
24.0 to 26.5%
Glutaraldehyde Solution, 25%
-5°C
111-30-8,7732-18-5
Glass Bottle
1 L
C5H8O2
MFCD00007025
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
pentanedial
3485
100.12
Certified
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Toxic by inhalation. Harmful if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction.
NFPA
Health: 3
Flammability: 1
Instability: 1

SIGNAL WORD: DANGER!

DOTInformation : DOT Class 6.1, : Poison

SDS
missing translation for 'documents'

missing translation for 'documents'