missing translation for 'onlineSavingsMsg'
Learn More

Glutaric Acid 99.0+%, TCI America™

$29.83 - $219.95

Chemical Identifiers

CAS 110-94-1
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD00004410
InChI Key JFCQEDHGNNZCLN-UHFFFAOYSA-N
Synonym glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di
PubChem CID 743
ChEBI CHEBI:17859
IUPAC Name pentanedioic acid
SMILES OC(=O)CCCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
G006925G
View Documents
TCI America
G006925G
25 g
Each for $29.83
Only null left
 
G0069500G
View Documents
TCI America
G0069500G
500 g
Each for $219.95
Only null left
 
Description

Description

Specifications

Chemical Identifiers

110-94-1
132.12
JFCQEDHGNNZCLN-UHFFFAOYSA-N
743
pentanedioic acid
C5H8O4
MFCD00004410
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di
CHEBI:17859
OC(=O)CCCC(O)=O

Specifications

110-94-1
White
C5H8O4
25 g
JFCQEDHGNNZCLN-UHFFFAOYSA-N
pentanedioic acid
743
132.12
Crystalline Powder
98°C
200°C
MFCD00004410
glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di
OC(=O)CCCC(O)=O
132.12
CHEBI:17859
≥99.0% (GC,T)
Glutaric Acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2560

RTECSNumber : MA3740000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'